Learn More
Percival Scientific Arabidopsis Growth Chamber, AR41L3C9, 37.2 cu. ft., SciWhite with red LEDs, 3 tiers, 7-44° C ±0.5°, IntellusUltraConnect controller, touchscreen interface, remote connectivity
SDP
Catalog No. NC3794513
Encompass_Preferred
The AR41L3C9 growth chamber is designed for growth of Arabidopsis and low light plant growth applications. SciWhite w/ red LEDs are dimmable from 10-100% output w/ up to 405 µmoles/m2/s irradiance @ 6". IntellusUltraConnect controller, touchscreen interface, remote connectivity. Interior/exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. Three white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.
Catalog No. NC3794513 Supplier Percival Scientific Supplier No. AR41L3C9
Add to Cart
Add to Cart
Request a Quote
The AR41L3C9 growth chamber is designed for growth of Arabidopsis and low light plant growth applications. SciWhite w/ red LEDs are dimmable from 10-100% output w/ up to 405 µmoles/m2/s irradiance @ 6". IntellusUltraConnect controller, touchscreen interface, remote connectivity. Interior/exterior of smooth white galvanized steel with foamed-in-place non-CFC insulation and a stainless-steel floor for corrosion resistance. Air circulation inside chamber is from a specifically designed adjustable air diffuser. Three white epoxy coated steel wire shelves adjustable in ½” increments. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; this also prolongs life of compressor, and eliminates risk of ice build up in coil. Top mounted refrigeration system allows easy access for maintenance and cleaning. Contact sales@percival-scientific.com for more options.