Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-arabinogalactan | 9036-66-2 | MFCD00062638 | 80.0% | C20H36O14 | 500 G
SDP

Catalog No. 5000282136
Encompass_Preferred

(+)-Arabinogalactan is a larch-wood-derived polysaccharide supplied as a powdered reagent with approximately 80% polyarabinogalactan content. It is intended for research use in glycobiology, biochemical assays, formulation studies, and analytical applications. Consult the product datasheet for batch-specific details and handling instructions.

  • Extracted from larch wood
  • Approximately 80% polyarabinogalactan content
  • Powdered form for reagent and formulation use
  • Typical applications: glycobiology and biochemical assays
  • CAS number 9036-66-2
  • Store at -20°C as recommended on datasheet

Catalog No. 50-002-82136 Supplier Medchemexpress LLC Supplier No. HYW350004500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Arabinogalactan is a larch-wood-derived polysaccharide supplied as a powdered reagent with approximately 80% polyarabinogalactan content. It is intended for research use in glycobiology, biochemical assays, formulation studies, and analytical applications. Consult the product datasheet for batch-specific details and handling instructions.

  • Extracted from larch wood
  • Approximately 80% polyarabinogalactan content
  • Powdered form for reagent and formulation use
  • Typical applications: glycobiology and biochemical assays
  • CAS number 9036-66-2
  • Store at -20°C as recommended on datasheet

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.