Learn More
Medchemexpress LLC Arabinogalactan from larch wood | 9036-66-2 | MFCD00062638 | 99.6% | 72-92 kDa | C20H36O14 | 10 G
SDP
Catalog No. 5000282563
Encompass_Preferred
Arabinogalactan from larch wood is a polysaccharide preparation (polyarabinogalactan) supplied as a powder for glycobiology and biochemical research. The material is reported as approximately 80% arabinogalactan by composition and has a manufacturer-reported assay of about 99.6%. Provided for research use only; follow recommended cold storage and handling conditions.
Catalog No. 50-002-82563 Supplier Medchemexpress LLC Supplier No. HYW35000410G
Add to Cart
Add to Cart
Arabinogalactan from larch wood is a polysaccharide preparation (polyarabinogalactan) supplied as a powder for glycobiology and biochemical research. The material is reported as approximately 80% arabinogalactan by composition and has a manufacturer-reported assay of about 99.6%. Provided for research use only; follow recommended cold storage and handling conditions.