Learn More
Medchemexpress LLC Arabinogalactan, from larch wood | 9036-66-2 | MFCD00062638 | 99.6% | Not specified - polymeric/variable (manufacturer does not list molecular weight) | Not specified by manufacturer - polymeric; representative formula C20H36O14 (repeating unit approx. (C5H8O4)n) | 25 G
SDP
Catalog No. 5000282121
Encompass_Preferred
Arabinogalactan from larch wood is a naturally derived polysaccharide supplied as an off-white to light yellow powder for glycobiology and biochemical research. It is water-soluble and commonly used as a carbohydrate reagent, stabilizer, or formulation component in in vitro assays; for research use only.
  • Natural polysaccharide extracted from larch wood.
  • Approx. 80% polyarabinogalactan composition.
  • Off-white to light yellow powder form, solid.
  • Water soluble; ~10 mg/mL (may require ultrasonic treatment).
  • High assay/purity (about 99.6%).
  • Stable when stored frozen; powder stable at -20°C for up to 3 years.
  • Intended for research use only.
Catalog No. 50-002-82121 Supplier Medchemexpress LLC Supplier No. HYW35000425G
Arabinogalactan from larch wood is a naturally derived polysaccharide supplied as an off-white to light yellow powder for glycobiology and biochemical research. It is water-soluble and commonly used as a carbohydrate reagent, stabilizer, or formulation component in in vitro assays; for research use only.
  • Natural polysaccharide extracted from larch wood.
  • Approx. 80% polyarabinogalactan composition.
  • Off-white to light yellow powder form, solid.
  • Water soluble; ~10 mg/mL (may require ultrasonic treatment).
  • High assay/purity (about 99.6%).
  • Stable when stored frozen; powder stable at -20°C for up to 3 years.
  • Intended for research use only.