Learn More
(+)-Arabinogalactan for use with Total Dietary Fiber Control Kit, TDF-C10, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| CAS | 9036-66-2 |
| Melting Point | >200°C |
| Packaging | Poly Bottle |
| MDL Number | MFCD00062638 |
| Quantity | 1 g |
| Synonym | Polyarabinogalactan |
| Physical Form | Lyophilized Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.