Learn More
Medchemexpress LLC Arabinose | 147-81-9 | 99.0% | 150.13 | 100 MG
SDP
Catalog No. 5000364722
Encompass_Preferred
Arabinose is a five-carbon sugar commonly found in plants. It alleviates immune disorders and inflammation by promoting balanced immune responses and reducing inflammation. It also serves as an intermediate in compound synthesis and is suitable for studies related to immune disorders and inflammation.
  • Five-carbon sugar found in plants
  • Alleviates immune disorders and inflammation
  • Promotes balanced immune responses
  • Reduces inflammation
  • Serves as an intermediate in compound synthesis
  • Applicable for studies in immune disorders and inflammation
  • Induces cytotoxicity, autophagy, and cell cycle arrest in breast cancer cells in vitro
  • Exerts neuromodulatory/antidepressant effects in neuroblastoma cells in vitro
Catalog No. 50-003-64722 Supplier Medchemexpress LLC Supplier No. 100MGHYN2353
Arabinose is a five-carbon sugar commonly found in plants. It alleviates immune disorders and inflammation by promoting balanced immune responses and reducing inflammation. It also serves as an intermediate in compound synthesis and is suitable for studies related to immune disorders and inflammation.
  • Five-carbon sugar found in plants
  • Alleviates immune disorders and inflammation
  • Promotes balanced immune responses
  • Reduces inflammation
  • Serves as an intermediate in compound synthesis
  • Applicable for studies in immune disorders and inflammation
  • Induces cytotoxicity, autophagy, and cell cycle arrest in breast cancer cells in vitro
  • Exerts neuromodulatory/antidepressant effects in neuroblastoma cells in vitro