Learn More
Medchemexpress LLC Arabinose-d | 77583-92-7 | 5 MG
SDP
Catalog No. 5000421404
Encompass_Preferred
Arabinose-d is a deuterium-labeled Arabinose, an endogenous metabolite. Stable heavy isotopes like hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers in quantifying during drug development. Deuteration is of interest due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-21404 Supplier Medchemexpress LLC Supplier No. HYN2353S15MG
Arabinose-d is a deuterium-labeled Arabinose, an endogenous metabolite. Stable heavy isotopes like hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers in quantifying during drug development. Deuteration is of interest due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.