Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arabinose-d | 77583-92-7 | >90.0% | 151.14 | C5H9DO5 | 10 MG
SDP

Catalog No. 5000289436
Encompass_Preferred

Arabinose-d is a deuterium-labeled form of the sugar arabinose supplied as a 10 mg stable isotope standard for research applications. It is intended for use as an internal standard, tracer, or analytical reference in metabolic and biochemical studies. For research use only.

  • Deuterium-labeled arabinose for isotope tracing and analytical studies.
  • Supplied as a 10 mg solid suitable for small-scale experiments.
  • CAS number 77583-92-7 for unambiguous identification.
  • Molecular formula C5H9DO5; molecular weight 151.14 g/mol.
  • Storage: store sealed at 4°C and protect from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only; not for clinical use.

Catalog No. 50-002-89436 Supplier Medchemexpress LLC Supplier No. HYN2353S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arabinose-d is a deuterium-labeled form of the sugar arabinose supplied as a 10 mg stable isotope standard for research applications. It is intended for use as an internal standard, tracer, or analytical reference in metabolic and biochemical studies. For research use only.

  • Deuterium-labeled arabinose for isotope tracing and analytical studies.
  • Supplied as a 10 mg solid suitable for small-scale experiments.
  • CAS number 77583-92-7 for unambiguous identification.
  • Molecular formula C5H9DO5; molecular weight 151.14 g/mol.
  • Storage: store sealed at 4°C and protect from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • Intended for research use only; not for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.