Learn More
System Biosciences LLC MINICIRRCLE ARABINOSE INDUCTION SOLUTIONArabinose induction solution (20%) = 20 mL
SDP
Catalog No. NC2669126
Encompass_Preferred
Induce minicircle production and removal of the vector backbone with this ready-to-add Arabinose Solution—use with our minicircle production strain ZYCY10P3S2T. Move your minicircle projects forward quickly and with confidence with SBI’s ready-to-add reagents for Minicircle Technology. Our Arabinose comes as a 20% solution, which is a 5x concentration for minicircle production.
Catalog No. NC2669126 Supplier System Biosciences LLC Supplier No. MN850A1
Induce minicircle production and removal of the vector backbone with this ready-to-add Arabinose Solution—use with our minicircle production strain ZYCY10P3S2T. Move your minicircle projects forward quickly and with confidence with SBI’s ready-to-add reagents for Minicircle Technology. Our Arabinose comes as a 20% solution, which is a 5x concentration for minicircle production.