Learn More
Cayman Chemical Arachidic Acid, Purity: ≥98%, CAS: 506-30-9, 100 mg
SDP
Catalog No. 502896884
Encompass_Preferred
A long-chain saturated fatty acid; inhibits N-formyl-Met-Leu-Phe-induced neutrophil aggregation at 5 μM
Catalog No. 50-289-6884 Supplier Cayman Chemical Supplier No. 9000339100 MG
A long-chain saturated fatty acid; inhibits N-formyl-Met-Leu-Phe-induced neutrophil aggregation at 5 μM