Learn More
Medchemexpress LLC Arachidic acid | 506-30-9 | 312.53 | 1 ML
SDP
Catalog No. 5000449241
Encompass_Preferred
Arachidic acid is an orally bioavailable membrane fluidity modulator and receptor/ion channel effector. It does not activate arachidonic acid-sensitive K+ channels in rat midbrain and hypothalamic neurons. Arachidic acid alleviates age-related spatial cognitive impairment and is applicable for research related to age-related spatial cognitive impairment.
  • Membrane fluidity modulator
  • Receptor/ion channel effector
  • Does not activate arachidonic acid-sensitive K+ channels
  • Alleviates age-related spatial cognitive impairment
  • Soluble in DMSO at 4.76 mg/mL
Catalog No. 50-004-49241 Supplier Medchemexpress LLC Supplier No. HYW00426010MM/1ML
Arachidic acid is an orally bioavailable membrane fluidity modulator and receptor/ion channel effector. It does not activate arachidonic acid-sensitive K+ channels in rat midbrain and hypothalamic neurons. Arachidic acid alleviates age-related spatial cognitive impairment and is applicable for research related to age-related spatial cognitive impairment.
  • Membrane fluidity modulator
  • Receptor/ion channel effector
  • Does not activate arachidonic acid-sensitive K+ channels
  • Alleviates age-related spatial cognitive impairment
  • Soluble in DMSO at 4.76 mg/mL