Learn More
Medchemexpress LLC Arachidic acid | 506-30-9 | 312.53 | 25 MG
SDP
Catalog No. 5000446681
Encompass_Preferred
Arachidic acid, also known as Eicosanoic acid, is an orally bioavailable membrane fluidity modulator and receptor/ion channel effector. It alleviates age-related spatial cognitive impairment and is applicable for research related to this condition. This compound does not activate arachidonic acid-sensitive K+ channels in rat midbrain and hypothalamic neurons.
  • Acts as an orally bioavailable membrane fluidity modulator
  • Functions as a receptor/ion channel effector
  • Shown to alleviate age-related spatial cognitive impairment
  • Applicable for research in neurodegenerative diseases
  • Stable as a powder for up to 3 years at -20°C and 2 years at 4°C
  • Exhibits solubility in DMSO at 4.76 mg/mL for in vitro applications
Catalog No. 50-004-46681 Supplier Medchemexpress LLC Supplier No. 25MGHYW004260R
Arachidic acid, also known as Eicosanoic acid, is an orally bioavailable membrane fluidity modulator and receptor/ion channel effector. It alleviates age-related spatial cognitive impairment and is applicable for research related to this condition. This compound does not activate arachidonic acid-sensitive K+ channels in rat midbrain and hypothalamic neurons.
  • Acts as an orally bioavailable membrane fluidity modulator
  • Functions as a receptor/ion channel effector
  • Shown to alleviate age-related spatial cognitive impairment
  • Applicable for research in neurodegenerative diseases
  • Stable as a powder for up to 3 years at -20°C and 2 years at 4°C
  • Exhibits solubility in DMSO at 4.76 mg/mL for in vitro applications