Learn More
Medchemexpress LLC Arachidic acid | 506-30-9 | 312.53 | 50 MG
SDP
Catalog No. 5000445611
Encompass_Preferred
Arachidic acid (Standard) is the analytical standard of Arachidic acid. This product is intended for research and analytical applications. Arachidic acid (Eicosanoic acid) is a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils. It can be conjugated with Chitosan oligosaccharide for anti-cancer drug delivery research.
  • Analytical standard for research and analytical applications
  • Long-chain saturated fatty acid
  • Found in human plasma phospholipid, fish, cannabis, and other plant oils
  • Can be conjugated with Chitosan oligosaccharide for anti-cancer drug delivery research
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
Catalog No. 50-004-45611 Supplier Medchemexpress LLC Supplier No. HYW004260R50MG
Arachidic acid (Standard) is the analytical standard of Arachidic acid. This product is intended for research and analytical applications. Arachidic acid (Eicosanoic acid) is a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils. It can be conjugated with Chitosan oligosaccharide for anti-cancer drug delivery research.
  • Analytical standard for research and analytical applications
  • Long-chain saturated fatty acid
  • Found in human plasma phospholipid, fish, cannabis, and other plant oils
  • Can be conjugated with Chitosan oligosaccharide for anti-cancer drug delivery research
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS