Learn More
Cayman Chemical Arachidic Acid, Purity: ≥98%, CAS: 506-30-9, 50 mg
SDP
Catalog No. 502892580
Encompass_Preferred
A long-chain saturated fatty acid; inhibits N-formyl-Met-Leu-Phe-induced neutrophil aggregation at 5 μM
Catalog No. 50-289-2580 Supplier Cayman Chemical Supplier No. 900033950 MG
A long-chain saturated fatty acid; inhibits N-formyl-Met-Leu-Phe-induced neutrophil aggregation at 5 μM