Learn More
Cayman Chemical ArachIdIc AcId-d2 100mg
SDP
Catalog No. 502939449
Encompass_Preferred
An internal standard for the quantification of arachidic acid by GC- or LC-MS
Catalog No. 50-293-9449 Supplier Cayman Chemical Supplier No. 28083100 MG
An internal standard for the quantification of arachidic acid by GC- or LC-MS