Learn More
Cayman Chemical Arachidic Acid-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 232600-70-3, 10 mg
SDP
Catalog No. 502938358
Encompass_Preferred
An internal standard for the quantification of arachidic acid by GC- or LC-MS
Catalog No. 50-293-8358 Supplier Cayman Chemical Supplier No. 2808310 MG
An internal standard for the quantification of arachidic acid by GC- or LC-MS