Learn More
Cayman Chemical Arachidic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 202480-70-4, 10 mg
SDP
Catalog No. 502937374
Encompass_Preferred
An internal standard for the quantification of arachidic acid by GC- or LC-MS
Catalog No. 50-293-7374 Supplier Cayman Chemical Supplier No. 2787410 MG
An internal standard for the quantification of arachidic acid by GC- or LC-MS