Learn More
Medchemexpress LLC Arachidic acid-d3 | 202480-70-4 | 315.55 | 5 MG
SDP
Catalog No. 5000445616
Encompass_Preferred
Arachidic acid-d3 is a deuterium-labeled version of Arachidic acid. Arachidic acid, also known as Eicosanoic acid, is a long-chain saturated fatty acid found in the human body within plasma phospholipids, fish, cannabis, and other plant oils. When conjugated with Chitosan oligosaccharide, Arachidic acid can be used in anti-cancer drug delivery research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-45616 Supplier Medchemexpress LLC Supplier No. HYW004260S25MG
Arachidic acid-d3 is a deuterium-labeled version of Arachidic acid. Arachidic acid, also known as Eicosanoic acid, is a long-chain saturated fatty acid found in the human body within plasma phospholipids, fish, cannabis, and other plant oils. When conjugated with Chitosan oligosaccharide, Arachidic acid can be used in anti-cancer drug delivery research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis through NMR, GC-MS, or LC-MS.
  • Deuterium substitution has the potential to influence the pharmacokinetic and metabolic profiles of drugs.