Learn More
Cayman Chemical ArachIdIc AcId-d3 25mg
SDP
Catalog No. 502939348
Encompass_Preferred
An internal standard for the quantification of arachidic acid by GC- or LC-MS
Catalog No. 50-293-9348 Supplier Cayman Chemical Supplier No. 2787425 MG
An internal standard for the quantification of arachidic acid by GC- or LC-MS