Learn More
Cayman Chemical Arachidic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 202480-70-4, 5 mg
SDP
Catalog No. 502939947
Encompass_Preferred
An internal standard for the quantification of arachidic acid by GC- or LC-MS
Catalog No. 50-293-9947 Supplier Cayman Chemical Supplier No. 278745 MG
An internal standard for the quantification of arachidic acid by GC- or LC-MS