Learn More
Medchemexpress LLC Arachidic acid-d39 | 39756-32-6 | 351.77 | 5 MG
SDP
Catalog No. 5000449152
Encompass_Preferred
Arachidic acid-d39 is the deuterium labeled Arachidic acid. Arachidic acid (Eicosanoic acid) is a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Used for anti-cancer drug delivery research when conjugated with chitosan oligosaccharide
Catalog No. 50-004-49152 Supplier Medchemexpress LLC Supplier No. HYW004260S15MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Arachidic acid-d39 is the deuterium labeled Arachidic acid. Arachidic acid (Eicosanoic acid) is a long-chain saturated fatty acid found in human plasma phospholipid, fish, cannabis, and other plant oils. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Used for anti-cancer drug delivery research when conjugated with chitosan oligosaccharide