Learn More
Cayman Chemical Arachidic Acid methyl ester, Purity: ≥98%, CAS: 1120-28-1, 1 g
SDP
Catalog No. 502936980
Encompass_Preferred
An esterified form of arachidic acid; a minor constituent of biodiesel generated from acid-catalyzed or SCW-SCM in situ transesterification of dry activated sludge or sludge oil, respectively
Catalog No. 50-293-6980 Supplier Cayman Chemical Supplier No. 267241 G
An esterified form of arachidic acid; a minor constituent of biodiesel generated from acid-catalyzed or SCW-SCM in situ transesterification of dry activated sludge or sludge oil, respectively