Learn More
Medchemexpress LLC Arachidic acid (Standard) | 506-30-9 | MFCD00002755 | 312.53 g/mol | 100 MG
SDP
Catalog No. 5000447468
Encompass_Preferred
Arachidic acid (Standard) is the analytical standard of Arachidic acid, also known as Eicosanoic acid. It is a long-chain saturated fatty acid naturally occurring in human plasma phospholipid, fish, cannabis, and various plant oils. This product is designed for research and analytical applications.
  • Intended for research and analytical applications
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Commonly applied in HPLC, GC, and MS analyses
  • Can be used for anti-cancer drug delivery research when conjugated with Chitosan oligosaccharide
Catalog No. 50-004-47468 Supplier Medchemexpress LLC Supplier No. HYW004260R100MG
Arachidic acid (Standard) is the analytical standard of Arachidic acid, also known as Eicosanoic acid. It is a long-chain saturated fatty acid naturally occurring in human plasma phospholipid, fish, cannabis, and various plant oils. This product is designed for research and analytical applications.
  • Intended for research and analytical applications
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Commonly applied in HPLC, GC, and MS analyses
  • Can be used for anti-cancer drug delivery research when conjugated with Chitosan oligosaccharide