Learn More
Cayman Chemical Arachidic Acid (sodium salt), Purity: ≥95%, CAS: 13257-34-6, 100 mg
SDP
Catalog No. 502985771
Encompass_Preferred
A long-chain saturated fatty acid; inhibits fMLP-induced rabbit neutrophil aggregation at 5 μM
Catalog No. 50-298-5771 Supplier Cayman Chemical Supplier No. 21906100 MG
A long-chain saturated fatty acid; inhibits fMLP-induced rabbit neutrophil aggregation at 5 μM