Learn More
Cayman Chemical ArachIdIc AcIdodIum salt 50mg
SDP
Catalog No. 502987170
Encompass_Preferred
A long-chain saturated fatty acid; inhibits fMLP-induced rabbit neutrophil aggregation at 5 μM
Catalog No. 50-298-7170 Supplier Cayman Chemical Supplier No. 2190650 MG
A long-chain saturated fatty acid; inhibits fMLP-induced rabbit neutrophil aggregation at 5 μM