Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arachidic Anhydride 98.0+%, TCI America™
SDP

Catalog No. A12331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12331G Supplier TCI America Supplier No. A12331G

Chemical Identifiers

CAS 55726-22-2
Molecular Formula C40H78O3
Molecular Weight (g/mol) 607.06
MDL Number MFCD00056235
InChI Key AUONNNVJUCSETH-UHFFFAOYSA-N
Synonym Eicosanoic Anhydride
PubChem CID 567077
IUPAC Name icosanoyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 77°C
Color Gray-White
Quantity 1 g
Formula Weight 607.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Arachidic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.