Learn More
Cayman Chemical Arachidonoyl Cyclopropylamide, Purity: ≥98%, CAS: 229021-64-1, 50 mg
SDP
Catalog No. 502894867
Encompass_Preferred
A potent and selective CB1 receptor agonist (Kis = 2.2 and 715 nM for CB1 and CB2, respectively); induces hypothermia in mice with the same efficacy as AEA in spite of its much higher affinity for the CB1 receptor and thus is a possible substrate for FAAH
Catalog No. 50-289-4867 Supplier Cayman Chemical Supplier No. 9105350 MG
A potent and selective CB1 receptor agonist (Kis = 2.2 and 715 nM for CB1 and CB2, respectively); induces hypothermia in mice with the same efficacy as AEA in spite of its much higher affinity for the CB1 receptor and thus is a possible substrate for FAAH