Learn More
Cayman Chemical ArachIdonIc ACD 1 2 3 4 513
SDP
Supplier:  Cayman Chemical 17336250 UG
Encompass_Preferred
An isotopically enriched form of a PUFA with carbon-13 occurring at positions 1, 2, 3, 4, and 5; used to study metabolic processes related to arachidonic acid by means of mass spectrometry
Catalog No. 50-296-6100