Learn More
Cayman Chemical Arachidonic Acid CYP450 Metabolite MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502982965
Encompass_Preferred
A mixture that contains (±)14(15)-DiHET (Item No. 51651), (±)11(12)-DiHET (Item No. 51511), (±)8(9)-DiHET (Item No. 51351), (±)5(6)-DiHET (Item No. 51211), 19(R)-HETE (Item No. 10007767), 20-HETE (Item No. 90030), (±)14(15)-EET (Item No. 50651), (±)11(12)-EET (Item No. 50511), and (±)8(9)-EET (Item No. 50351)
Catalog No. 50-298-2965 Supplier Cayman Chemical Supplier No. 206651 EA
A mixture that contains (±)14(15)-DiHET (Item No. 51651), (±)11(12)-DiHET (Item No. 51511), (±)8(9)-DiHET (Item No. 51351), (±)5(6)-DiHET (Item No. 51211), 19(R)-HETE (Item No. 10007767), 20-HETE (Item No. 90030), (±)14(15)-EET (Item No. 50651), (±)11(12)-EET (Item No. 50511), and (±)8(9)-EET (Item No. 50351)