Learn More
Cayman Chemical Arachidonic Acid methyl ester, Purity: ≥98%, CAS: 2566-89-4, 100 mg
SDP
Catalog No. 502895692
Encompass_Preferred
An esterified form of arachidonic acid; commonly used as a reference standard for the quantification of arachidonic acid in biological samples and as a source of exogenous arachidonic acid in cells and in vivo
Catalog No. 50-289-5692 Supplier Cayman Chemical Supplier No. 90014100 MG
An esterified form of arachidonic acid; commonly used as a reference standard for the quantification of arachidonic acid in biological samples and as a source of exogenous arachidonic acid in cells and in vivo