Learn More
Cayman Chemical ArachIdonIc ACD mthyl STR 50mg
SDP
Catalog No. 502897018
Encompass_Preferred
An esterified form of arachidonic acid; commonly used as a reference standard for the quantification of arachidonic acid in biological samples and as a source of exogenous arachidonic acid in cells and in vivo
Catalog No. 50-289-7018 Supplier Cayman Chemical Supplier No. 9001450 MG
An esterified form of arachidonic acid; commonly used as a reference standard for the quantification of arachidonic acid in biological samples and as a source of exogenous arachidonic acid in cells and in vivo