Learn More
Cayman Chemical Arachidonic Acid Oxylipin MaxSpec LC-MS Mixture, 1 ea
SDP
Catalog No. 502984492
Encompass_Preferred
A mixture that contains 5(S)-HETE , 5-OxoETE , 8(S)-HETE , 9(R)-HETE , 11(S)-HETE , 12(S)-HETE , 12-OxoETE , 15(S)-HETE, and 15-OxoETE.
Catalog No. 50-298-4492 Supplier Cayman Chemical Supplier No. 206661 EA
A mixture that contains 5(S)-HETE , 5-OxoETE , 8(S)-HETE , 9(R)-HETE , 11(S)-HETE , 12(S)-HETE , 12-OxoETE , 15(S)-HETE, and 15-OxoETE.