Learn More
Cayman Chemical ArachIdonIc ACD sulfonatS
SDP
Supplier:  Cayman Chemical 9001886500 UG
Encompass_Preferred
An α-sulphonated version of arachidonic acid that may possess improved lipophilicity
Catalog No. 50-289-5359