Learn More
Cayman Chemical ArachIdonIc ACDd8 methyl S 1mg
SDP
Catalog No. 502919485
Encompass_Preferred
An internal standard for the quantification of arachidonic acid methyl ester by GC- or LC-MS
This item is not returnable. View return policy
Catalog No. 50-291-9485 Supplier Cayman Chemical Supplier No. 3900141 MG
Add to Cart
Add to Cart
An internal standard for the quantification of arachidonic acid methyl ester by GC- or LC-MS