Learn More
TARGETMOL CHEMICALS INC ARACHIDONIC ACID 100MG
SDP
Catalog No. 502576803
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Arachidonic acid (Immunocytophyte) is an unsaturated essential fatty acid. It is found in animal and human fat as well as in the liver brain and glandular organs and is a constituent of animal phosphatides. It is formed by the synthesis from dietary linoleic acid and is a precursor in the biosynthesis of prostaglandins thromboxanes and leukotrienes. purity: 99%
Catalog No. 50-257-6803 Supplier TARGETMOL CHEMICALS INC Supplier No. T4129100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Arachidonic acid (Immunocytophyte) is an unsaturated essential fatty acid. It is found in animal and human fat as well as in the liver brain and glandular organs and is a constituent of animal phosphatides. It is formed by the synthesis from dietary linoleic acid and is a precursor in the biosynthesis of prostaglandins thromboxanes and leukotrienes. purity: 99%