Learn More
Medchemexpress LLC HY-109590 10mg Medchemexpress, Arachidonic acid CAS:506-32-1 Purity:>98%
SDP
Catalog No. 501968712
Encompass_Preferred
Medchemexpress, HY-109590 10mg Arachidonic acid CAS:506-32-1 Arachidonic acid is an essential fatty acid and a major constituent of biomembranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8712 Supplier Medchemexpress LLC Supplier No. HY10959010MG
Medchemexpress, HY-109590 10mg Arachidonic acid CAS:506-32-1 Arachidonic acid is an essential fatty acid and a major constituent of biomembranes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.