Learn More
Medchemexpress LLC Arachidonic acid-13C4 (immunocytophyt-13C4) | 2692624-40-9 | 95.0% | 308.44 | 10 UMOL
SDP
Catalog No. 5000344467
Encompass_Preferred
Arachidonic acid-13C4 is a 13C labeled Arachidonic acid. Arachidonic acid (Immunocytophyt) is a polyunsaturated omega-6 fatty acid and a major constituent of biomembranes. It also acts as the substrate for various lipid mediators, such as prostaglandins (PGs). Arachidonic acid improves cognitive response and cardiovascular function.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-44467 Supplier Medchemexpress LLC Supplier No. HY109590S3100UG
Arachidonic acid-13C4 is a 13C labeled Arachidonic acid. Arachidonic acid (Immunocytophyt) is a polyunsaturated omega-6 fatty acid and a major constituent of biomembranes. It also acts as the substrate for various lipid mediators, such as prostaglandins (PGs). Arachidonic acid improves cognitive response and cardiovascular function.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS