Learn More
Enzo Life Sciences Arachidonic acid (20:4, n-6) (1g). CAS: 506-32-1
SDP
Supplier:  Enzo Life Sciences BMLFA0031000
Encompass_Preferred
Precursor to a large family of eicosanoids. Acts as a second messenger independent of metabolism. Retrograde messenger in long-term potentiation in the nervous system. Inhibits ras-GAP. Activator for several protein kinase C (PKC) isotypes. Alternative name: 5Z,8Z,11Z,14Z-Eicosatetraenoic acid. Purity: ≥99% (TLC). Solubility: 100mg/ml soluble in DMSO or dimethyl formamide; soluble in 100% ethanol (50mg/ml); sparingly soluble in basic aqueous buffers (1.7mg/ml). To prevent oxidation, the solvent should be purged with an inert gas. Long Term Storage: -20°C.
Catalog No. 50-201-0794