Learn More
Cayman Chemical Arachidonic Acid, 506-32-1, 250 mg
SDP
Supplier:  Cayman Chemical 90010250
Encompass_Preferred
Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals
Catalog No. NC0279213