Learn More
MilliporeSigma ARACHIDONIC ACID
Catalog No. 502303612
Encompass
Arachidonic acid; > /=99% by TLC; Colorless oil; EMD; Calbiochem; 100mg; cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid; F.W. 304.5; C20H32O2 M.P. 169 to 171 deg.C (336.2 to 339.8 deg.F); CAS: C
Catalog No. 50-230-3612 Supplier MilliporeSigma Supplier No. 181198100MG
Arachidonic acid; > /=99% by TLC; Colorless oil; EMD; Calbiochem; 100mg; cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid; F.W. 304.5; C20H32O2 M.P. 169 to 171 deg.C (336.2 to 339.8 deg.F); CAS: C