Learn More
Cayman Chemical Arachidonic Acid (peroxide free), 506-32-1, 50 mg
SDP
Supplier:  Cayman Chemical 90010.150
Encompass_Preferred
Molecular Formula C20H32O2, Purity ≥98%, Formula Weight 304.5, Synonyms: 5Z,8Z,11Z,14Z-eicosatetraenoic acid, AA (peroxide free); Free arachidonic acid is a transient, critical substrate for the biosynthesis of eicosanoid second messengers. Arachidonic acid oxidizes spontaneously on exposure to room air, yielding a complex mixture of fatty acid peroxides. Peroxide-free arachidonic acid is obtained after removal of the peroxides by silicic acid chromatography, followed by immediate addition of the antioxidant BHT (2,6-Di-tert-butyl-4-hydroxytoluene). Arachidonic acid is also available without BHT (Catalog No. 90010).
Catalog No. NC0206100