Learn More
Cayman Chemical Arachidonic Acid, 506-32-1, 100 mg
SDP
Supplier:  Cayman Chemical 90010100
Encompass_Preferred
Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals
Catalog No. NC9267260