Learn More
Cayman Chemical Arachidonic Acid, Purity: ≥98%, CAS: 506-32-1, 500 mg
SDP
Catalog No. 502892393
Encompass_Preferred
Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals
Catalog No. 50-289-2393 Supplier Cayman Chemical Supplier No. 90010500 MG
Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals