Learn More
Cayman Chemical Arachidonic Acid, Purity: ≥98%, CAS: 506-32-1, 50 mg
SDP
Catalog No. 502892514
Encompass_Preferred
Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals
Catalog No. 50-289-2514 Supplier Cayman Chemical Supplier No. 9001050 MG
Keystone essential fatty acid that is converted by COX, LO, and epoxygenase enzymes into more than 150 potent metabolites in species ranging from fungi to plants to mammals