Learn More
Biomatik Corporation Arachidonic Acid (AA) ELISA Kit
Supplier:  Biomatik Corporation EKU02560
Encompass_Preferred
Arachidonic Acid (AA) ELISA Kit for detecting AA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 2.47-200ug/ml. Sensitivity: 0.88ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: ARA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9118