Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arachidonic acid-biotin | 1217901-28-4 | 95% | 100 UG
SDP

Catalog No. 5000342660
Encompass_Preferred

Arachidonic acid-biotin is a biotin-labeled arachidonic acid primarily used as a laboratory chemical. This product has a purity of 95% and is identified by CAS number 1217901-28-4.

  • Biotin-labeled arachidonic acid
  • Purity of 95%
  • CAS number 1217901-28-4
  • Ideal for research use in laboratory settings
  • Store as a solution at -20°C for up to 2 years
  • Appears as a colorless to light yellow liquid
  • Molecular formula: C35H58N4O3S
  • Molecular weight: 614.93

Catalog No. 50-003-42660 Supplier Medchemexpress LLC Supplier No. HY127049100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Arachidonic acid-biotin is a biotin-labeled arachidonic acid primarily used as a laboratory chemical. This product has a purity of 95% and is identified by CAS number 1217901-28-4.

  • Biotin-labeled arachidonic acid
  • Purity of 95%
  • CAS number 1217901-28-4
  • Ideal for research use in laboratory settings
  • Store as a solution at -20°C for up to 2 years
  • Appears as a colorless to light yellow liquid
  • Molecular formula: C35H58N4O3S
  • Molecular weight: 614.93

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.