Learn More
Cayman Chemical Arachidonic Acid-d11, 1 mg
SDP
Supplier:  Cayman Chemical 100067581
Encompass_Preferred
An internal standard for the quantification of arachidonic acid by GC- or LC-MS
Catalog No. NC0910529