Learn More
Medchemexpress LLC Arachidonic acid-d5 | 123167-26-0 | ≥98.0% | 309.50 | 100 MG
SDP
Catalog No. 5000344670
Encompass_Preferred
Arachidonic acid-d5 is the deuterium labeled Arachidonic acid. Arachidonic acid is an essential fatty acid and a major constituent of biomembranes. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Essential fatty acid
  • Major constituent of biomembranes
  • For research use only
Catalog No. 50-003-44670 Supplier Medchemexpress LLC Supplier No. HY109590S11MG
Arachidonic acid-d5 is the deuterium labeled Arachidonic acid. Arachidonic acid is an essential fatty acid and a major constituent of biomembranes. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Essential fatty acid
  • Major constituent of biomembranes
  • For research use only