Learn More
Cayman Chemical Arachidonic Acid-d8, Purity: ≥99% deuterated forms (d1-d8), CAS: 69254-37-1, 1 mg
SDP
Catalog No. 502919482
Encompass_Preferred
An internal standard for the quantification of arachidonic acid by GC- or LC-MS
Catalog No. 50-291-9482 Supplier Cayman Chemical Supplier No. 3900101 MG
An internal standard for the quantification of arachidonic acid by GC- or LC-MS