Learn More
Cayman Chemical Arachidonic Acid ethyl ester, Purity: ≥98%, CAS: 1808-26-0, 500 mg
SDP
Catalog No. 502889561
Encompass_Preferred
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure
Catalog No. 50-288-9561 Supplier Cayman Chemical Supplier No. 10008200500 MG
An esterified form of arachidonic acid; commonly used as a source of exogenous arachidonic acid in vivo; has been found in the meconium of newborns suspected of fetal alcohol exposure